missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPOX Antibody (36B10), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59438AF594
This item is not returnable.
View return policy
Description
CPOX Monoclonal antibody specifically detects CPOX in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CPOX | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Coprogen oxidase, coproporphyrinogen oxidase, coproporphyrinogenase, coproporphyrinogen-III oxidase, mitochondrial, COX, CPO, CPXcoproporphyrinogen oxidase (coproporphyria, harderoporphyria), EC 1.3.3.3, HCP | |
| Recombinant human CPOX (111-454aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1371 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 36B10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction