missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPO Antibody (OTI1C1), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72386PECY55
This item is not returnable.
View return policy
Description
CPO Monoclonal specifically detects CPO in Human samples. It is validated for Flow Cytometry.
Specifications
| CPO | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CPO | |
| Full length recombinant protein of human CPO (NP_775100) produced in HEK293T cells. | |
| 0.1 mL | |
| 130749.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1C1 | |
| Flow Cytometry | |
| carboxypeptidase O, EC 3.4.17, EC 3.4.17.-, MGC138281, MGC138283 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction