missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPNE6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13782-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.Specifications
| CPNE6 | |
| Polyclonal | |
| Unconjugated | |
| CPNE6 | |
| Copine 6, Copine VI, copine VI (neuronal), CPNE6, N COPINE, Neuronal copine | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12891, 691478, 9362 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95741, Q9Z140 | |
| CPNE6 | |
| CPNE6 Fusion Protein Ag4789 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction