missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPNE6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527621
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (99%), Rat (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.
Specifications
| CPNE6 | |
| Polyclonal | |
| Unconjugated | |
| CPNE6 | |
| AU067659; BB076446; copine 6; copine VI; copine VI (neuronal); copine-6; Cpne6; MGC137714 protein; N-copine; neuronal copine; neuronal-copine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95741 | |
| CPNE6 | |
| Recombinant protein encompassing a sequence within the center region of human CPNE6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction