Learn More
Invitrogen™ CPNE6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556806
Description
Immunogen sequence: DMSDPEMGWV PEPPTMTLGA SRVELRVSCH GLLDRDTLTK PHPCVLLKLY SDEQWVEVER TEVLRSCSSP VFSRVLALEY FFEEKQPLQF HVFDAEDGAT SPRNDTFLGS TECTLGQIVS QTKVTKPLLL KNGKTAGKST ITIVAEEVSG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.
Specifications
| CPNE6 | |
| Polyclonal | |
| Unconjugated | |
| CPNE6 | |
| AU067659; BB076446; copine 6; copine VI; copine VI (neuronal); copine-6; Cpne6; MGC137714 protein; N-copine; neuronal copine; neuronal-copine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95741 | |
| CPNE6 | |
| Recombinant protein corresponding to Human CPNE6. Recombinant protein control fragment (Product #RP-88929). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.