missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101498
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CPN1.
CPN1 is a plasma metallo-protease that cleaves basic amino acids from the C terminal of peptides and proteins. The protein is important in the regulation of peptides like kinins and anaphylatoxins, and has also been known as kininase-1 and anaphylatoxin inactivator. This protein is a tetramer comprised of two identical regulatory subunits and two identical catalytic subunits.
Specifications
| CPN1 | |
| Polyclonal | |
| Unconjugated | |
| CPN1 | |
| 0610011F20Rik; ACBP; Anaphylatoxin inactivator; arginine carboxypeptidase; carboxypeptidase K; carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; carboxypeptidase N small subunit; carboxypeptidase N subunit 1; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; cb1037; CPN; CPN1; fa99g08; FLJ40792; kininase I; kininase-1; Lysine carboxypeptidase; plasma carboxypeptidase B; SCPN; serum carboxypeptidase N; Unknown (protein for MGC:128810); wu:fa99g08; zgc:77485 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1369, 93721 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15169, Q9JJN5 | |
| CPN1 | |
| A synthesized peptide derived from human CPN1(Accession P15169), corresponding to amino acid residues I419-A458. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction