missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21222-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| CPLX4 | |
| Polyclonal | |
| Unconjugated | |
| CPLX4 | |
| complexin 4, Complexin IV, CPLX4, CPX IV, CPXIV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225644, 339302, 361342 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z7G2, Q80WM3 | |
| CPLX4 | |
| CPLX4 Fusion Protein Ag15661 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction