missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00305CL1
This item is not returnable.
View return policy
Description
CPLX3 Polyclonal antibody specifically detects CPLX3 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| CPLX3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 594855 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| complexin 3, Complexin III, complexin-3, CPX III, CPXIII, CPX-III, DKFZp434E1719, FLJ00167, FLJ00250, FLJ13993 | |
| Produced in rabbits immunized with purified, recombinant Human CPLX3 (Uniprot#: Q8WVH0; Met1-Lys154) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction