missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPLX2 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
| Antigen | CPLX2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CPLX2 |
| Gene Accession No. | P84086, P84087, Q6PUV4 |
| Gene Alias | 921-L; AI413745; AW492120; complexin 2; complexin II; Complexin-2; CPLX2; CPX II; CPX2; CPX-2; CPX-II; DKFZp547D155; Hfb1; LOW QUALITY PROTEIN: complexin-2; MGC138492; synaphin; synaphin 1; synaphin-1; wu:fc27b12 |
| Gene Symbols | CPLX2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?