missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPLX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119049
This item is not returnable.
View return policy
Description
CPLX1 Polyclonal Antibody for Western Blot
CPLX1 encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release.
Specifications
| CPLX1 | |
| Polyclonal | |
| Unconjugated | |
| Cplx1 | |
| 921-S; complexin 1; complexin I; Complexin-1; Cplx1; CPX I; CPX1; CPX-I; synaphin 2; synaphin-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10815, 12889, 64832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O14810, P63040, P63041 | |
| Cplx1 | |
| Recombinant protein Complexin 1. The antigen corresponds to amino acid range 1-134 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction