missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPI17 alpha Antibody (4H10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04336H
This item is not returnable.
View return policy
Description
CPI17 alpha Monoclonal specifically detects CPI17 alpha in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CPI17 alpha | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PPP1R14A | |
| Recombinant human PPP1R14A (1-147aa) purified from E. coli | |
| 0.1 mL | |
| Phospho Specific, Protein Phosphatase | |
| 94274 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4H10 | |
| Western Blot, ELISA | |
| 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human PPP1R14A mAb, clone 4H10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPP1R14A protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction