missing translation for 'onlineSavingsMsg'
Learn More

CPI-637, MedChemExpress

Product Code. 16414346
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414346

Brand: Medchem Express HY100482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CPI-637 is a selective and potent CBP/EP300 bromodomain inhibitor with IC50 values of 0.03 μM, 0.051 μM and 11.0 μM for CBP, EP300 and BRD4 BD-1, respectively, and an EC50 of 0.3 μM for CBP.

CPI-637 is a selective and potent CBP/EP300 bromodomain inhibitor with IC50 values of 0.03 μM, 0.051 μM and 11.0 μM for CBP, EP300 and BRD4 BD-1, respectively, and an EC50 of 0.3 μM for CBP.

Specifications

Chemical Name or Material CPI-637
Physical Form Solid
CAS 1884712-47-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H22N6O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 9.62 mg/mL (24.89 mM; Need ultrasonic)
SMILES O=C1NC2=CC=CC(C3=CC4=C(N(C)N=C4C5=CN(C)N=C5)C=C3)=C2N[C@H](C)C1
Molecular Weight (g/mol) 386.45
Formula Weight 386.45
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.