missing translation for 'onlineSavingsMsg'
Learn More

CPI-444, TRC

Product Code. 30358084
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358084 25 mg 25mg
30351460 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358084 Supplier Toronto Research Chemicals Supplier No. TRCC78145525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-7-(5-methylfuran-2-yl)-3-((6-(((tetrahydrofuran-3-yl)oxy)methyl)pyridin-2-yl)methyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-5-amine
CAS 1202402-40-1
Molecular Formula C20H21N7O3
Quantity 25 mg
Synonym CPI-444, Triazolo-pyramidine Derivative
InChI Formula InChI=1S/C20H21N7O3/c1-12-5-6-16(30-12)17-18-19(24-20(21)23-17)27(26-25-18)9-13-3-2-4-14(22-13)10-29-15-7-8-28-11-15/h2-6,15H,7-11H2,1H3,(H2,21,23,24)/t15-/m0/s1
SMILES Cc1oc(cc1)c2nc(N)nc3c2nnn3Cc4cccc(CO[C@H]5CCOC5)n4
IUPAC Name 7-(5-methylfuran-2-yl)-3-[[6-[[(3S)-oxolan-3-yl]oxymethyl]pyridin-2-yl]methyl]triazolo[4,5-d]pyrimidin-5-amine
Molecular Weight (g/mol) 407.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.