missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112012
This item is not returnable.
View return policy
Description
CPF1 Polyclonal Antibody for Western Blot, ICC/IF
Clustered Regularly Interspaced Short Palindromic Repeats from Prevotella and Francisella 1 or CRISPR/Cpf1 is a DNA-editing technology analogous to the CRISPR/Cas9 system. Cpf1 is an RNA-guided endonuclease of a class II CRISPR/Cas system. This acquired immune mechanism is found in Prevotella and Francisella bacteria.
Specifications
| CPF1 | |
| Polyclonal | |
| Unconjugated | |
| Cpf1 locus; CRISPR; CRISPR-Cas | |
| Recombinant protein encompassing a sequence within the C-terminus region of Lachnospiraceae bacterium ND2006 CPF1 protein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.12 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction