missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPEB3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CPEB3 |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632992
|
Novus Biologicals
NBP2-92754-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612592
|
Novus Biologicals
NBP2-92754-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPEB3 Polyclonal antibody specifically detects CPEB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CPEB3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 22849 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CPE-binding protein 3, cytoplasmic polyadenylation element binding protein 3, cytoplasmic polyadenylation element-binding protein 3, hCPEB-3, KIAA0940CPE-BP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-400 of human CPEB3 (NP_055727.3). TFSPQIGLAQTQHHQQPPPPAPAPQPAQPAQPPQAQPPQQRRSPASPSQAPYAQRSAAAAYGHQPIMTSKPSSSSAVAAAAAAAAASSASSSWNTHQSVNAAWSAPSNPWGGLQAGRDPRRAVGVGVGVGVGVPSPLNPISPLKKPFSSNVIAPPKFPRAAPLTSKSWMEDNAFRTDNGNNLLPFQDRSRPYDTFNLHSLENSLMDMIRTDHEPLKGKHYPPSGPPMSFADIMWRNHFAGRMGINFHHPGT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title