missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPEB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567245
This item is not returnable.
View return policy
Description
Immunogen sequence: NTLLPLQVRSS LQLPAWGSDS LQDSWCTAAG TSRIDQDRSR MYDSLNMHS Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The protein encoded by this gene is highly similar to cytoplasmic polyadenylation element binding protein (CPEB), an mRNA-binding protein that regulates cytoplasmic polyadenylation of mRNA as a trans factor in oogenesis and spermatogenesis. Studies of the similar gene in mice suggested a possible role of this protein in transcriptionally inactive haploid spermatids. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| CPEB2 | |
| Polyclonal | |
| Unconjugated | |
| CPEB2 | |
| CPE-BP2; CPEB-2; hCPEB-2; CPE-binding protein 2; cytoplasmic polyadenylation element-binding protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z5Q1 | |
| CPEB2 | |
| Recombit Human CPEB2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction