missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPEB1 Recombinant Rabbit Monoclonal Antibody (23GB3495), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552563
This item is not returnable.
View return policy
Description
This gene encodes a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the nuclear membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the nuclear membrane and inhibit 5-lipoxygenase activation. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
| CPEB1 | |
| Recombinant Monoclonal | |
| 1.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0C279, P70166, Q9BZB8 | |
| CPEB1 | |
| A synthetic peptide of human CPEB1 (265-325AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3495 | |
| Unconjugated | |
| CPEB1 | |
| AU024112; CEBP; Cpeb; CPEB1; CPEB-1; CPE-binding protein 1; CPE-BP1; cytoplasmic polyadenylation element binding protein 1; cytoplasmic polyadenylation element-binding protein 1; FLJ13203; h-CEBP; h-CPEB; hCPEB-1; mCPEB; mCPEB-1; MGC34136; MGC60106 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12877, 293056, 64506 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction