missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CPEB1 Polyclonal antibody specifically detects CPEB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | CPEB1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CPEBCEBP, CPE-binding protein 1, CPE-BP1, cytoplasmic polyadenylation element binding protein 1, cytoplasmic polyadenylation element-binding protein 1, FLJ13203, h-CEBP, hCPEB-1, MGC34136, MGC60106 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human CPEB1. The immunogen is located within amino acids 50 - 100 of CPEB1. Amino Acid Squence: LPDFQDSEETVTSR |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?