Learn More
Cole-Parmer™ CPC™ Polysulfone Coupling Plug
Click to view available options
:
3/4 in. OD
Description
- With their locking feature, these couplings guard against accidental disconnect and spills.
- To assemble, place the insert in the coupling body and twist to lock.
- An audible click ensures your connection.
- All couplings are made from USP Class VI-compliant polysulfone with platinum-cured silicone seals.
- Available in 3/4in. size, these fittings allow quick fill or empty of bioprocessing bags.
Specifications
Specifications
| Description | Quick-Disconnect x Barb |
| Diameter (English) Inner | 3/4 in. |
| Material | Polysulfone |
| Packaging Quantity | 10/Pk. |
| Sterilization Method | Autoclave, Ethylene Oxide or Gamma |
| Product Type | Coupling Plug |