Learn More
CPC™ Plastic Quick-Disconnect Fittings
Description
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables
CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.
- Material: Medical-grade ABS, polycarbonate and polysulfone
- Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
- Fow Rate: Cv∽5.5 (79.25) max.
- Ergonomic thumb latch
- Audible 'click' when connected
- USP Class VI materials
- Can be sterilized by autoclave, EtO, e-beam, or gamma
- Coupling halves can be rotated when connected
- Tapered barb
- BSE/TSE free materials
- Cell culture media packaging and transfer lines
- Harvest and collection applications
- Buffer solutions
- Tube sets
- Urology drainage/collection systems
- Hypo/hyperthermia blankets
Specifications
Specifications
| Certifications/Compliance | USP Class VI, ADCF |
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 0.25 in. |
| Diameter (Metric) Inner | 6.4 mm |
| Material | Polycarbonate |
| Overall Length (English) | 1.1 in. |
| Overall Length (Metric) | 28 mm |
| For Use With | 6.4mm |
| For Use With (Application) | Cell culture media packaging and transfer lines, harvest and collection applications |
| Min. Operating Temperature (English) | -40°F |
| Show More |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.