Learn More
CP110 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-12780-100UL
Description
The CP110 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CP110. This antibody recognizes human antigen. The CP110 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| CP110 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Immunofluorescence | |
| CoraLite 594 | |
| 100 μL | |
| 9738 | |
| IgG |