missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CP054 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524693
This item is not returnable.
View return policy
Description
CP054 Polyclonal Antibody for Western Blot, Flow
Specifications
| CP054 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6UWD8 | |
| C16orf54 | |
| KLH conjugated synthetic peptide between 170-198 amino acids from the C-terminal region of human CP054. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| C16orf54 | |
| C16orf54; chromosome 16 open reading frame 54; transmembrane protein C16orf54; UNQ9389/PRO34280 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 283897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction