Learn More
Chrompack CP-Sil 19 CB, 15 m, 0.32 mm, 0.25 µm, 7 inch Cage
Brand:  Chrompack CP8542
Encompass_Preferred
CP-Sil 19 CB, 15 m, 0.32 mm, 0.25 µm, 7 inch Cage
Product Code. 15513484
  • £546.75 / Each