missing translation for 'onlineSavingsMsg'
Learn More

CP 96345, Tocris Bioscience™

Product Code. 13431994
Encompass
Click to view available options
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NK1 antagonist

Potent and selective non-peptide NK1 receptor antagonist. Attenuates substance P-induced salivary response and inhibits neurogenic inflammation in vivo.

Specifications

CAS 132746-60-2
Molecular Formula C28H32N2O
Quantity 50mg
Synonym unii-w22ila2i52, chembl16192, 2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-cis-2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-n-2-methoxyphenyl methyl-2-diphenylmethyl-1-azabicyclo 2.2.2 octan-3-amine, 1-azabicyclo 2.2.2 octan-3-amine, 2-diphenylmethyl-n-2-methoxyphenyl methyl-, 2s-cis, 2s,3s-2-benzhydryl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, d03sxn, 2-benzhydryl-1-aza-bicyclo 2.2.2 oct-3-yl-2-methoxy-benzyl-amine
InChI Key FLNYLINBEZROPL-NSOVKSMOSA-N
SMILES COC1=CC=CC=C1CNC2C3CCN(C2C(C4=CC=CC=C4)C5=CC=CC=C5)CC3
IUPAC Name (2S,3S)-2-benzhydryl-N-[(2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine
Molecular Weight (g/mol) 412.577
PubChem CID 104943
Formula Weight 412.57
Percent Purity >98%
Chemical Name or Material CP 96345
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'