missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 945598 hydrochloride, Tocris Bioscience™
CB1 receptor antagonist
£206.95 - £810.08
Chemical Identifiers
CAS | 686347-12-6 |
---|---|
Molecular Formula | C25H26Cl3N7O |
Molecular Weight (g/mol) | 546.881 |
InChI Key | KPYUQCJBZGQHPL-UHFFFAOYSA-N |
Synonym | otenabant hydrochloride, cp 945598 hydrochloride, otenabant hcl, cp945598 hydrochloride, otenabant hydrochloride usan, otenabant usan/inn, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide hydrochloride, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide monohydrochloride |
PubChem CID | 16223963 |
IUPAC Name | 1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide;hydrochloride |
SMILES | CCNC1(CCN(CC1)C2=NC=NC3=C2N=C(N3C4=CC=C(C=C4)Cl)C5=CC=CC=C5Cl)C(=O)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18033832
|
Tocris Bioscience
4236/10 |
10 mg |
£206.95
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18004432
|
Tocris Bioscience
4236/50 |
50 mg |
£810.08
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective, high affinity CB1 antagonist (Ki values are 0.7 and 0.12 nM in binding and functional assays respectively). Displays low affinity for CB2 receptors (Ki = 7600 nM).Chemical Identifiers
686347-12-6 | |
546.881 | |
otenabant hydrochloride, cp 945598 hydrochloride, otenabant hcl, cp945598 hydrochloride, otenabant hydrochloride usan, otenabant usan/inn, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide hydrochloride, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide monohydrochloride | |
1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide;hydrochloride |
C25H26Cl3N7O | |
KPYUQCJBZGQHPL-UHFFFAOYSA-N | |
16223963 | |
CCNC1(CCN(CC1)C2=NC=NC3=C2N=C(N3C4=CC=C(C=C4)Cl)C5=CC=CC=C5Cl)C(=O)N.Cl |
Specifications
686347-12-6 | |
otenabant hydrochloride, cp 945598 hydrochloride, otenabant hcl, cp945598 hydrochloride, otenabant hydrochloride usan, otenabant usan/inn, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide hydrochloride, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide monohydrochloride | |
CCNC1(CCN(CC1)C2=NC=NC3=C2N=C(N3C4=CC=C(C=C4)Cl)C5=CC=CC=C5Cl)C(=O)N.Cl | |
546.881 | |
546.88 | |
CP 945598 hydrochloride |
C25H26Cl3N7O | |
KPYUQCJBZGQHPL-UHFFFAOYSA-N | |
1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide;hydrochloride | |
16223963 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C