missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 94253 hydrochloride, Tocris Bioscience™
Potent and selective 5-HT1B agonist
£183.58 - £697.20
Chemical Identifiers
CAS | 845861-39-4 |
---|---|
Molecular Formula | C15H20ClN3O |
Molecular Weight (g/mol) | 293.795 |
InChI Key | PIIOXKQIZCVXMD-UHFFFAOYSA-N |
Synonym | cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride |
PubChem CID | 11652258 |
IUPAC Name | 5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride |
SMILES | CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30001233
|
Tocris Bioscience
1317/10 |
10 mg |
£183.58
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18044412
|
Tocris Bioscience
1317/50 |
50 mg |
£697.20
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, selective 5-HT1B agonist (Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively). Centrally active upon systemic administration in vivo.Chemical Identifiers
845861-39-4 | |
293.795 | |
cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride | |
5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride |
C15H20ClN3O | |
PIIOXKQIZCVXMD-UHFFFAOYSA-N | |
11652258 | |
CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl |
Specifications
845861-39-4 | |
cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride | |
CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl | |
293.795 | |
293.8 | |
CP 94253 hydrochloride |
C15H20ClN3O | |
PIIOXKQIZCVXMD-UHFFFAOYSA-N | |
5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride | |
11652258 | |
>98% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature