missing translation for 'onlineSavingsMsg'
Learn More

CP-724714, MedChemExpress

Product Code. 16486128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486128

Brand: Medchem Express HY14674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CP-724714 is a potent, selective and orally active ErbB2 (HER2) tyrosine kinase inhibitor, with an IC50 of 10 nM. CP-724714 displays a marked selectivity against EGFR kinase (IC50=6400 nM). CP-724714 potently inhibits ErbB2 receptor autophosphorylation in intact cells. Antitumor activities.

CP-724714 is a potent, selective and orally active ErbB2 (HER2) tyrosine kinase inhibitor, with an IC50 of 10 nM. CP-724714 displays a marked selectivity against EGFR kinase (IC50=6400 nM). CP-724714 potently inhibits ErbB2 receptor autophosphorylation in intact cells. Antitumor activities.

Specifications

Chemical Name or Material CP-724714
CAS 383432-38-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H27N5O3
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (106.49 mM)
SMILES O=C(COC)NC/C=C/C1=CC2=C(N=CN=C2C=C1)NC3=CC=C(C(C)=C3)OC4=CC=C(N=C4)C.[(E)]
Molecular Weight (g/mol) 469.53
Formula Weight 469.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.