missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP-640186 hydrochloride, MedChemExpress
Description
CP-640186 hydrochloride is a potent and cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 53 nM and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2 respectively.
Specifications
Specifications
| Chemical Name or Material | CP-640186 hydrochloride |
| CAS | 591778-70-0 |
| Color | Pink |
| Purity Grade Notes | Research |
| Molecular Formula | C30H36ClN3O3 |
| Quantity | 100 mg |
| Solubility Information | H2O : 50 mg/mL (95.77 mM; Need ultrasonic) ∣DMSO : ≥ 48 mg/mL (91.94 mM) |
| SMILES | O=C([C@H]1CN(C2CCN(C(C3=C(C=CC=C4)C4=CC5=C3C=CC=C5)=O)CC2)CCC1)N6CCOCC6.Cl |
| Molecular Weight (g/mol) | 522.08 |
| Formula Weight | 522.08 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?