missing translation for 'onlineSavingsMsg'
Learn More

CP-466722, MedChemExpress

Product Code. 16487057
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487057 5 mg 5mg
16444348 10 mg 10mg
16400677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487057 Supplier Medchem Express Supplier No. HY11002
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CP-466722 is a rapidly reversible inhibitor of ATM, with an IC50 of 4.1 μM, and has no effects on PI3K or closely related PI3K-like protein kinase (PIKK) family members.

CP-466722 is a rapidly reversible inhibitor of ATM, with an IC50 of 4.1 μM, and has no effects on PI3K or closely related PI3K-like protein kinase (PIKK) family members.

Specifications

Chemical Name or Material CP-466722
CAS 1080622-86-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H15N7O2
Quantity 5 mg
Solubility Information DMSO : 1 mg/mL (2.86 mM; Need ultrasonic)
SMILES NC1=NC(C2=NC=CC=C2)=NN1C3=C4C=C(OC)C(OC)=CC4=NC=N3
Molecular Weight (g/mol) 349.35
Formula Weight 349.35
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.