missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CP 43

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CP 43 is a potent TAO kinase (TAOK) inhibitor (IC50 values are 11 nM/L and 15 nM/L for TAOK1 and TAOK2, respectively). Inhibits TAOK phosphorylation and activation of JNK in vitro. Prolongs duration of mitosis and increases mitotic cell death in the SKBR3 breast cancer cell line. Also reduces tau phopshorylation in cortical neurons at sites associated with neurodegeneration in human tauopathies.
Specifications
Specifications
| Chemical Name or Material | N-(2-Oxo-2-((1,2,3,4-tetrahydronaphthalen-1-yl)amino)ethyl)-[1,1′-biphenyl]-4-carboxamide |
| CAS | 850467-66-2 |
| Quantity | 50 mg |
| Molecular Formula | C25H24N2O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction