missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 339818 hydrochloride, Tocris Bioscience™
Non-peptide, potent KV1.3 channel blocker
Chemical Identifiers
CAS | 478341-55-8 |
---|---|
Molecular Formula | C20H23ClN2 |
Molecular Weight (g/mol) | 326.868 |
InChI Key | HDAMNMJLKSDNJF-UHFFFAOYSA-N |
Synonym | n-1-benzylquinolin-4-ylidene butan-1-amine hydrochloride |
PubChem CID | 71433722 |
IUPAC Name | 1-benzyl-N-butylquinolin-4-imine;hydrochloride |
SMILES | CCCCN=C1C=CN(C2=CC=CC=C12)CC3=CC=CC=C3.Cl |
Description
Potent, non-peptide KV1.3 channel antagonist that preferentially binds to the C-type inactivated state of the channel (IC50 ∽ 200 nM) . Inhibits KV1.4 with an IC50 of ∽ 300 nM. Selective over KV1.1, KV1.2, KV1.5, KV1.6, KV3.1-4, and KV4.2.Chemical Identifiers
478341-55-8 | |
326.868 | |
n-1-benzylquinolin-4-ylidene butan-1-amine hydrochloride | |
1-benzyl-N-butylquinolin-4-imine;hydrochloride |
C20H23ClN2 | |
HDAMNMJLKSDNJF-UHFFFAOYSA-N | |
71433722 | |
CCCCN=C1C=CN(C2=CC=CC=C12)CC3=CC=CC=C3.Cl |
Specifications
478341-55-8 | |
n-1-benzylquinolin-4-ylidene butan-1-amine hydrochloride | |
CCCCN=C1C=CN(C2=CC=CC=C12)CC3=CC=CC=C3.Cl | |
326.868 | |
340.89 | |
CP 339818 hydrochloride |
C20H23ClN2 | |
HDAMNMJLKSDNJF-UHFFFAOYSA-N | |
1-benzyl-N-butylquinolin-4-imine;hydrochloride | |
71433722 | |
>99% |
Safety and Handling
Recommended Storage : Store at Room Temperature