missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 31398 dihydrochloride, Tocris Bioscience™
p53-stabilizing agent
Chemical Identifiers
| CAS | 1217195-61-3 |
|---|---|
| Molecular Formula | C22H29ClN4O2 |
| Molecular Weight (g/mol) | 416.95 |
| InChI Key | HUVADZBUMSMSIQ-IVKCLRODSA-N |
| Synonym | cp-31398 dihydrochloride hydrate |
| PubChem CID | 131698900 |
| IUPAC Name | N-[2-[(E)-2-(4-methoxyphenyl)ethenyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine;hydrate;hydrochloride |
| SMILES | CN(C)CCCNC1=NC(=NC2=CC=CC=C21)C=CC3=CC=C(C=C3)OC.O.Cl |
Description
p53 stabilizing agent. Stabilizes the active conformation of p53 and promotes p53 activity in cancer cell lines with mutant or wild-type p53. Inhibits growth of small human tumor xenografts in vivo.Chemical Identifiers
| 1217195-61-3 | |
| 416.95 | |
| cp-31398 dihydrochloride hydrate | |
| N-[2-[(E)-2-(4-methoxyphenyl)ethenyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine;hydrate;hydrochloride |
| C22H29ClN4O2 | |
| HUVADZBUMSMSIQ-IVKCLRODSA-N | |
| 131698900 | |
| CN(C)CCCNC1=NC(=NC2=CC=CC=C21)C=CC3=CC=C(C=C3)OC.O.Cl |
Specifications
| 1217195-61-3 | |
| cp-31398 dihydrochloride hydrate | |
| CN(C)CCCNC1=NC(=NC2=CC=CC=C21)C=CC3=CC=C(C=C3)OC.O.Cl | |
| 416.95 | |
| 435.39 | |
| CP 31398 dihydrochloride |
| C22H29ClN4O2 | |
| HUVADZBUMSMSIQ-IVKCLRODSA-N | |
| N-[2-[(E)-2-(4-methoxyphenyl)ethenyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine;hydrate;hydrochloride | |
| 131698900 | |
| >99% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title