missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coxsackievirus A6 VP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112007
This item is not returnable.
View return policy
Description
Coxsackievirus A6 VP3 Polyclonal Antibody for Western Blot
Coxsackievirus A6 (CVA6) is a human enterovirus associated with herpangina in infants. VP1, VP2, and VP3 all have a compact core structure consisting of a classical, beta-sandwich, jelly-roll fold, which forms the bulk of the capsid and each of these protein subunits also have several loops that decorate the outer surface of the capsid.
Specifications
| Coxsackievirus A6 VP3 | |
| Polyclonal | |
| Unconjugated | |
| Coxsackievirus | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Coxsackievirus A6 VP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.63 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu