missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coxsackievirus A6 VP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112001
This item is not returnable.
View return policy
Description
Coxsackievirus A6 VP1 Polyclonal Antibody for Western Blot, IHC (F), ICC/IF
Coxsackievirus A6 (CVA6) is a human enterovirus associated with herpangina in infants. VP1, VP2, and VP3 all have a compact core structure consisting of a classical, beta-sandwich, jelly-roll fold, which forms the bulk of the capsid and each of these protein subunits also have several loops that decorate the outer surface of the capsid.
Specifications
| Coxsackievirus A6 VP1 | |
| Polyclonal | |
| Unconjugated | |
| Coxsackievirus | |
| Recombinant protein corresponding to Coxsackievirus A6 VP1 protein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 1.06 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction