missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX8C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103395
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytochrome c Oxidase 8C.
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water and ATP. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. The COX8 (cytochrome c oxidase subunit VIII) subunits are nuclear and have muscle and non-muscle-specific isoforms. COX8 exists as three isoforms: COX8a, a liver and heart isoform, Cox8b, a heart-specific isoform, and Cox8c, whose expression pattern has yet to be elucidated. All three Cox8 isoforms exists as components of the COX complex and play an important role in electron transport. This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
| COX8C | |
| Polyclonal | |
| Unconjugated | |
| COX8C | |
| 1700007F21Rik; COX VIII-3; COX8-3; Cox8c; COXVIII-3; cytochrome c oxidase polypeptide 8; Cytochrome c oxidase polypeptide 8 isoform 3; cytochrome c oxidase polypeptide VIII; Cytochrome c oxidase polypeptide VIII isoform 3; Cytochrome c oxidase subunit 8-3; cytochrome c oxidase subunit 8C; cytochrome c oxidase subunit 8C, mitochondrial; cytochrome c oxidase subunit VIII; cytochrome c oxidase subunit VIIIC | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 341947, 75483 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6H666, Q7Z4L0 | |
| COX8C | |
| A synthesized peptide derived from human COX8C(Accession Q7Z4L0), corresponding to amino acid residues R5-F55. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction