missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX7B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550190
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total COX7B2 protein.
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water and ATP. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7b (cytochrome c oxidase subunit VIIb polypeptide) and COX7b2 (cytochrome c oxidase subunit VIIb polypeptide 2) are 80 and 81 amino acid proteins, respectively, which exist as components of the COX complex, therefore playing an important role in electron transport. A rare polymorphism in the COX7b2 gene at codon 26 may be linked to nasopharyngeal carcinoma (NPC), the most common head and neck cancer in southern China.
Specifications
| COX7B2 | |
| Polyclonal | |
| Unconjugated | |
| COX7B2 | |
| 4930503B16Rik; COX7B2; Cytochrome c oxidase polypeptide VIIb2; cytochrome c oxidase subunit 7B2; cytochrome c oxidase subunit 7B2, mitochondrial; cytochrome c oxidase subunit VIIb2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170712 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8TF08 | |
| COX7B2 | |
| Full length fusion protein of Human COX7B2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction