missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599807
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total COX7B.
Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex. This nuclear gene encodes subunit VIIb, which is highly similar to bovine COX VIIb protein and is found in all tissues. This gene may have several pseudogenes on chromosomes 1, 2, 20 and 22, respectively.
Specifications
| COX7B | |
| Polyclonal | |
| Unconjugated | |
| COX7B | |
| 1100001F07Rik; 1110004F07Rik; APLCC; C80563; COX7B; Cytochrome c oxidase polypeptide VIIb; cytochrome c oxidase subunit 7B; cytochrome c oxidase subunit 7B, mitochondrial; cytochrome c oxidase subunit VIIb; cytochrome-c oxidase chain VIIb; LSDMCA2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1349, 66142 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24311, P56393 | |
| COX7B | |
| A synthesized peptide derived from human COX7B(Accession P24311), corresponding to amino acid residues K6-V56. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction