missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX7A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549922
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total COX7A3 / COX7A2P2 protein.
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
| COX7A2 | |
| Polyclonal | |
| Unconjugated | |
| COX7A2 | |
| Cox7a2; Cox7a3; COX7AL; COX7AL1; COX7AL2; COXVIIAL; COXVIIa-L; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; cytochrome c oxidase subunit 7A2; cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa 2; cytochrome c oxidase subunit VIIa 3; cytochrome c oxidase subunit VIIa polypeptide 2; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); cytochrome c oxidase subunit VIIaL; cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase, subunit 7a 3; cytochrome c oxidase, subunit VIIa 2; hepatic cytochrome-c oxidase chain VIIa; VIIAL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12866, 1347, 1348, 29507 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60397, P14406, P35171, P48771 | |
| COX7A2, COX7A2P2 | |
| Synthesized peptide derived from internal of human COX7S/A2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction