missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX7A2L Polyclonal Antibody

COX7A2L Polyclonal Antibody
Brand: Invitrogen PA599808
This item is not returnable.
View return policy
Description
COX7A2L Polyclonal Antibody for Western Blot
Specifications
| COX7A2L | |
| Polyclonal | |
| Unconjugated | |
| Cox7a2l | |
| COX7A2L; COX7AR; COX7a-related protein; COX7RP; cytochrome c oxidase subunit 7A2 like; cytochrome c oxidase subunit 7A-related protein, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 like; cytochrome c oxidase subunit VIIa polypeptide 2-like; cytochrome c oxidase subunit VIIa-related protein; cytochrome c oxidase subunit VII-related protein; EB1; estrogen receptor binding CpG island; Scaf1; SIG81; SIG-81; Silg81; Silica-induced gene 81 protein; supercomplex assembly factor I | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20463, 298762, 9167 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14548, Q61387 | |
| Cox7a2l | |
| A synthesized peptide derived from human COX7A2L(Accession O14548), corresponding to amino acid residues P35-M85. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction