missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Positive Samples: HeLa Immunogen sequence: MLRNLLALRQ IGQRTISTAS RRHFKNKVPE KQKLFQEDDE IPLYLKGGVA DALLYRATMI LTVGGTAYAI YELAVASFPK KQE.
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
Specifications
| Antigen | COX7A2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | COX7A2P2 |
| Gene Accession No. | O60397, P14406 |
| Gene Alias | Cox7a2; Cox7a3; COX7AL; COX7AL1; COX7AL2; COXVIIAL; COXVIIa-L; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; cytochrome c oxidase subunit 7A2; cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa 2; cytochrome c oxidase subunit VIIa 3; cytochrome c oxidase subunit VIIa polypeptide 2; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); cytochrome c oxidase subunit VIIaL; cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase, subunit 7a 3; cytochrome c oxidase, subunit VIIa 2; hepatic cytochrome-c oxidase chain VIIa; VIIAL |
| Gene Symbols | COX7A2, COX7A2P2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
