missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COX6C Recombinant Rabbit Monoclonal Antibody (K01_1Q11)

Product Code. 30830415
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830415 50 μL
30825408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830415 Supplier Invitrogen™ Supplier No. MA564995
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DFEEMRKAGI FQSVK.

Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q11
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene COX6C
Gene Accession No. P09669, P11951, Q9CPQ1
Gene Alias COVIc; Cox6c; Cox6c2; COX-VIc; cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; cytochrome c oxidase subunit 6C; Cytochrome c oxidase subunit 6C-2; cytochrome c oxidase subunit VIc; cytochrome c oxidase subunit VIc preprotein; cytochrome c oxidase, subunit VIc; cytochrome oxidase subunit VIc; EC 1.9.3.1
Gene Symbols COX6C
Host Species Rabbit
Immunogen A synthetic peptide of human Cytochrome C Oxidase subunit Vic (aa 61-75).
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12864, 1345, 54322
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.