missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX6C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553141
This item is not returnable.
View return policy
Description
Immunogen sequence: EPRKKAYADF YRNYDVMKDF EDMRKTGT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
Specifications
| COX6C | |
| Polyclonal | |
| Unconjugated | |
| COX6C | |
| COVIc; Cox6c; Cox6c2; COX-VIc; cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; cytochrome c oxidase subunit 6C; Cytochrome c oxidase subunit 6C-2; cytochrome c oxidase subunit VIc; cytochrome c oxidase subunit VIc preprotein; cytochrome c oxidase, subunit VIc; cytochrome oxidase subunit VIc; EC 1.9.3.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09669 | |
| COX6C | |
| Recombinant protein corresponding to Human COX6C. Recombinant protein control fragment (Product #RP-102541). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction