missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6B2 Antibody (OTI1F5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72379CL1
This item is not returnable.
View return policy
Description
COX6B2 Monoclonal antibody specifically detects COX6B2 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| COX6B2 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cancer/testis antigen 59, COX VIb-2, COXVIB2, CT59MGC119094, cytochrome c oxidase subunit 6B2, Cytochrome c oxidase subunit VIb isoform 2, cytochrome c oxidase subunit VIb polypeptide 2 (testis), cytochrome c oxidase subunit VIb, testes specific, cytochrome c oxidase subunit VIb, testes-specific, Cytochrome c oxidase subunit VIb, testis-specific isoform, FLJ32865, FLJ46422 | |
| Full length human recombinant protein of human COX6B2 (NP_653214) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1F5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 125965 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction