missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11425-1-AP-150UL
This item is not returnable.
View return policy
Description
Connects the two COX monomers into the physiological dimeric form.Specifications
| COX6B1 | |
| Polyclonal | |
| Unconjugated | |
| COX6B1 | |
| COX VIb 1, COX6B, COX6B1, COXG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110323, 1340, 688869 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14854, P56391, P80430 | |
| Cox6b1 | |
| COX6B1 Fusion Protein Ag1984 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction