missing translation for 'onlineSavingsMsg'
Learn More

COX6B1 Rabbit anti-Human, Clone: 7T0J5, Novus Biologicals™

Product Code. 18353326 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353326 100 μg 100µL
18366794 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18353326 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP316098100UL

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Monoclonal Antibody

COX6B1 Monoclonal antibody specifically detects COX6B1 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen COX6B1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7T0J5
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias COX VIb-1, COX6B, COXG, COXVIb1, cytochrome c oxidase subunit 6B1, cytochrome c oxidase subunit Vib, Cytochrome c oxidase subunit VIb isoform 1, cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-86 of human COX6B1 (P14854).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1340
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.