Learn More
COX6B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552070
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EDMETKIKNY KTAPFDSRFP NQNQTRNCWQ NYLDFHRCQK AMTAKGGDIS VCEWYQRVYQ SLCPTSWVTD WDEQRAEGTF P Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Connects the two COX monomers into the physiological dimeric form.Specifications
COX6B1 | |
Polyclonal | |
Unconjugated | |
COX6B1 | |
2010000G05Rik; COX VIb-1; Cox6b; COX6B1; COXG; COXVIb1; cytochrome c oxidase subunit 6B1; cytochrome c oxidase subunit VIb; Cytochrome c oxidase subunit VIb isoform 1; cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous); cytochrome c oxidase, subunit VIb polypeptide 1; LOC688869; similar to cytochrome c oxidase, subunit VIb polypeptide 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1340 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14854 | |
COX6B1 | |
Recombinant protein corresponding to Human COX6B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only