missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX6A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596256
This item is not returnable.
View return policy
Description
Immunogen sequence: SSGAHGEEGS ARMWKTLTFF VALPGVAVSM LNVYLKSHHG EHERPEFIAY PHLRIRTKPF PWGDGNHTLF HNPHVNPLPT GYEDE; Positive Samples: A-549, MCF7, Mouse kidney, Mouse heart, Mouse liver, Rat liver, Rat brain; Cellular Location: Mitochondrion inner membrane.
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
| COX6A1 | |
| Polyclonal | |
| Unconjugated | |
| Cox6a1 | |
| CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12861, 1337, 25282 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10818, P12074, P43024 | |
| Cox6a1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-109 of human COX6A1 (NP_0043642). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction