missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX5B Polyclonal Antibody

COX5B Polyclonal Antibody
Brand: Invitrogen PA5103384
This item is not returnable.
View return policy
Description
COX5B Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| COX5B | |
| Polyclonal | |
| Unconjugated | |
| COX5B | |
| COX5B; COXVB; cytochrome c oxidase polypeptide Vb; cytochrome c oxidase polypeptide VB, mitochondrial; cytochrome c oxidase polypeptide VIa; cytochrome c oxidase precursor (EC 1.9.3.1); cytochrome c oxidase subunit 5B; cytochrome c oxidase subunit 5B, mitochondrial; cytochrome c oxidase subunit Vb; cytochrome c oxidase subunit VIA* | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12859, 1329, 94194 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10606, P12075, P19536 | |
| COX5B | |
| A synthesized peptide derived from human COX5B(Accession P10606), corresponding to amino acid residues A14-V64. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction