missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556963
This item is not returnable.
View return policy
Description
Immunogen sequence: ASGGGVPTDE EQATGLEREI MLAAKKGLDP YNVLAPKGAS GTREDPNLVP SISNKRIVGC ICEEDNTSVV WFWLHKGEAQ RCPRCGAHYK LVP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. Found in the inner mitochondrial membrane, COX5 is the heme A-containing chain of the oxidase family that converts one molecule of oxygen and four molecules of hydrogen to two molecules of water. Two isoforms of COX5 exist, COX5a and COX5b. When oxygen levels within the cell are high, transcription of COX5a (the aerobic isoform) is up-regulated as the rate of cellular respiration increases. Conversely, when oxygen levels are low, COX5b (the hypoxic isoform) transcription increases and functions to maximize the turnover rate of the COX apoenzyme.
Specifications
| COX5B | |
| Polyclonal | |
| Unconjugated | |
| COX5B | |
| COX5B; COXVB; cytochrome c oxidase polypeptide Vb; cytochrome c oxidase polypeptide VB, mitochondrial; cytochrome c oxidase polypeptide VIa; cytochrome c oxidase precursor (EC 1.9.3.1); cytochrome c oxidase subunit 5B; cytochrome c oxidase subunit 5B, mitochondrial; cytochrome c oxidase subunit Vb; cytochrome c oxidase subunit VIA* | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1329 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10606 | |
| COX5B | |
| Recombinant protein corresponding to Human COX5B. Recombinant protein control fragment (Product #RP-95792). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion