missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX5b Antibody (029), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90313CL1
This item is not returnable.
View return policy
Description
COX5b Monoclonal antibody specifically detects COX5b in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| COX5b | |
| Monoclonal | |
| CoraFluor 1 | |
| COXVB, Cytochrome c oxidase polypeptide Vb, cytochrome c oxidase polypeptide VB, mitochondrial, cytochrome c oxidase subunit 5B, mitochondrial, cytochrome c oxidase subunit Vb | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human COX5b (Uniprot#: P10606; Ala32-His129). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 1329 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 029 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction